missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human PILR-alpha Fc Chimera Protein

Product Code. 18775396
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18775396

Brand: R&D Systems™ 10802PR050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Measured by its binding ability in a functional ELISA. When Recombinant Human PANP Fc Chimera (Catalog # 7920-PN ) is coated at 5.00 μg/mL (100 μL/well), Recombinant Human PILR-alpha Fc Chimera binds with an ED50 of 0.150- 0.900 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UKJ1.3
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 29992
Molecular Weight (g/mol) M.W.-observed: 57-64 kDa, under reducing conditions, M.W.-theroretical: 46 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human PILR-alpha protein Human PILR-alpha (Thr25-Thr196) (N-terminus) Accession # Q9UKJ1.3 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias Cell surface receptor FDF03, FDF03, inhibitory receptor PILRalpha, Inhibitory receptor PILR-alpha, paired immunoglobin-like receptor alpha, paired immunoglobin-like type 2 receptor alpha, paired immunoglobulin-like receptor alpha, paired immunoglobulin-like type 2 receptor alpha, PILRA, PILRalpha
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein PILR-alpha
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.