missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Oncostatin M (OSM), Animal-Free Protein

Product Code. 18796065
Click to view available options
Quantity:
50 μg
500 μg
Unit Size:
500µg
50µg
This item is not returnable. View return policy

Product Code. 18796065

Brand: R&D Systems™ Qk0490050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
No significant difference between EC50 of reference and test lots
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P13725.2
For Use With (Application) Bioactivity
Formulation Lyophilized from acetonitrile/TFA
Gene ID (Entrez) 5008
Molecular Weight (g/mol) M.W. (Observed): Monomeric Oncostatin M (OSM) protein only, M.W. (Theroretical): 22 kDa
Quantity 50 μg
Source E. coli-derived human Oncostatin M/OSM protein
Storage Requirements Store lyophilized protein between -20°C to -80°C until the date of expiry. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias MGC20461, oncostatin M, oncostatin-M
Conjugate Unconjugated
Reconstitution Resuspend in water at >100 μg/mL, prepare single use aliquots, add carrier protein if desired.
Protein Oncostatin M/OSM
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.