missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-2 R beta Fc Chimera Protein

Product Code. 18769496
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18769496

Brand: R&D Systems™ 109192B050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Measured by its ability to inhibit Recombinant Human IL-15 (Catalog # 247-ILB ) dependent proliferation of MO7e human megakaryocytic leukemic cells. The ED50 for this effect is 1.75-14.0 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P14784.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 3560
Molecular Weight (g/mol) M.W.-observed: 62 - 68 kDa, under reducing conditions., M.W.-theroretical: 51 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human IL-2 R beta protein Human IL-2 R beta (Ala27-Asp239) (C-terminus) Accession # P14784.1 IEGRMD Human IgG1 (Pro100-Lys330) (N-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CD122, CD122 antigen, high affinity IL-2 receptor beta subunit, High affinity IL-2 receptor subunit beta, IL-15 R beta, IL-2 receptor subunit beta, IL2R beta, IL-2 R subunit beta, IL2RB, IL-2 RB, interleukin 15 receptor, beta, interleukin 2 receptor, beta, interleukin-2 receptor subunit beta, P70-75, p75
Conjugate Unconjugated
Reconstitution Reconstitute at 250 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL-2 R beta
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.