missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ SPINeasy™ DNA Kit for Tissue and Bacteria

Product Code. 17361549 Shop All MP Biomedicals Products
Click to view available options
Quantity:
50 Preps
Unit Size:
50 reactions
This item is not returnable. View return policy

Product Code. 17361549

Brand: MP Biomedicals™ 116532050

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Fast and easy isolation of high-quality genomic DNA from animal tissues, blood, cultured cells, and bacteria. Lysing Matrix A for thorough and complete lysis with FastPrep Instruments. High-performance gDNA extraction kit based on silica membrane spin column technology.

  • SPINeasy DNA Kit for Tissue and Bacteria is a high-performance genomic DNA (gDNA) extraction kit based on silica membrane spin column technology.
  • The kit enables quick isolation of gDNA from various animal tissues, blood, cultured cells, and bacteria, typically in less than 30 minutes.
  • The use of our specially formulated Lysis Buffer GD and Lysing Matrix A in combination with FastPrep instruments from MP Biomedicals enables highly efficient lysis of various tissue types within seconds.
  • The provided Column GD coupled with specially formulated kit buffers are designed to deliver gDNA of high yield and purity with fewer steps, suitable for downstream applications such as restriction digestion, PCR, and NGS.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at Room Temperature, Dry Environment
Isolation Technology Spin Column
Sample Type Blood, Tissue, Bacteria, Insects, Swabs
Includes 50 x 2 mL Lysing Matrix A, 60 mL Lysis Buffer GD, 30 mL Wash Buffer GD1, 6 mL Wash Buffer GD2, 10 mL Elution Buffer GD, 50 x Column GD with collection tubes
Quantity 50 Preps
compliance-icons
Product Identifier
  • SPINeasy Plasmid Miniprep Kit, 50 preps
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Sodium Dodecyl Sulphate, sodium hydroxide

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.