missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ IND 1316

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
IND 1316 is an activator of AMP-activated protein kinase (AMPK). In vitro, it activates AMPK in a dose-dependent manner and induces different MAPK pathways (p38 and JNK). In vivo, it reduces the neuropathological symptoms in two animal models of Huntington's Disease (nematodes and mice). It shows very low toxicity even at high concentrations (>200 μM). This compound is orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 3-[3-(Phenylmethoxy)phenyl]-1 H-indole-2-carboxylic acid |
| CAS | 1888473-12-8 |
| Quantity | 50 mg |
| Target | AMPK Activators |
| Molecular Formula | C22H17NO3 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction