missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ERGi-USU
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
ERGi-USU is a high affinity Rio Kinase 2 (RIOK2) inhibitor (Kd = 200 nM). Exhibits >6-fold selectivity for RIOK2 over a panel of 456 kinases. Inhibits ERG protein and potently inhibits growth of ERG-positive VCaP prostate cancer cells in vitro (IC50 = 169 nM) but not ERG-negative cell lines. Also inhibits growth of VCaP xenografts in mice.
Specifications
Specifications
| Chemical Name or Material | 1-[2-(2-Thiazolyl)diazenyl]-2-naphthalenol |
| CAS | 1147-56-4 |
| Quantity | 50 mg |
| Molecular Formula | C13H9N3OS |
| Formula Weight | 255.3 |
| Purity | 99% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction