missing translation for 'onlineSavingsMsg'
Learn More

(1S,3R)-ACPD, Tocris Bioscience™

Product Code. 30001072
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001072

Brand: Tocris Bioscience 0284/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Group I and group II mGlu agonist; active isomer of (±)-trans-ACPD (Cat. No. 0187)

Active isomer of (±)-trans-ACPD . Agonist at both group I and II mGlu receptors (EC50 values are 5, 15, 42 and 60 μM at mGluR2, mGluR5, mGluR1 and mGluR6 respectively).

NPEC-caged-(1S,3R)-ACPD, (±)-trans-ACPD and cis-ACPD also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111900-32-4
Molecular Formula C7H11NO4
Molecular Weight (g/mol) 173.168
InChI Key YFYNOWXBIBKGHB-FBCQKBJTSA-N
Synonym 1s,3r-1-aminocyclopentane-1,3-dicarboxylic acid, 1s,3r-acpd, trans-acpd, 1-amino-1,3-dicarboxycyclopentane, trans-+/--acpd, chembl34453, 1s,3s-acpd, 1-aminocyclopentane-1s,3r-dicarboxylic acid, 1s,3r-1-acpd
PubChem CID 104766
IUPAC Name (1S,3R)-1-aminocyclopentane-1,3-dicarboxylic acid
SMILES C1CC(CC1C(=O)O)(C(=O)O)N

Specifications

Quantity 50 mg
Formula Weight 173.17
Percent Purity >98%
Chemical Name or Material (1S, 3R)-ACPD
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.