missing translation for 'onlineSavingsMsg'
Learn More

5-Vinylbenzo[d][1,3]dioxole (stabilized with TBC), TRC

Product Code. 30453568
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453568 50 mg 50mg
30451712 500 mg 500mg
30451689 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30453568 Supplier Toronto Research Chemicals Supplier No. TRCV37520350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7315-32-4
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.16
IUPAC Name 5-ethenyl-1,3-benzodioxole
SMILES C=Cc1ccc2OCOc2c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H8O2/c1-2-7-3-4-8-9(5-7)11-6-10-8/h2-5H,1,6H2
Formula Weight 148.0524
Chemical Name or Material 5-Vinylbenzo[d][1,3]dioxole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.