missing translation for 'onlineSavingsMsg'
Learn More

5-Vinyl-2-norbornene, 98%, stabilized

Product Code. 10503151 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10503151 500 mL Glass bottle 500mL
10357630 25 mL Glass bottle 25mL
10105543 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10503151 Supplier Thermo Scientific Acros Supplier No. 164865000
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3048-64-4 | C9H12 | 120.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3048-64-4
Molecular Formula C9H12
Molecular Weight (g/mol) 120.19
MDL Number MFCD00080691
InChI Key INYHZQLKOKTDAI-UHFFFAOYSA-N
Synonym 5-vinyl-2-norbornene, 2-vinylnorbornene, vinylnorbornene, 5-vinylnorbornene, 2-vinyl-5-norbornene, 2-norbornene, 5-vinyl, 5-vinylnorborn-2-ene, bicyclo 2.2.1 hept-2-ene, 5-ethenyl, vinylnorbornene van, 5-vinylbicyclo 2.2.1 hept-2-ene
PubChem CID 18273
IUPAC Name 5-ethenylbicyclo[2.2.1]hept-2-ene
SMILES C=CC1CC2CC1C=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -80°C
Boiling Point 141°C
Flash Point 27°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4800 to 1.482
Quantity 500 mL
Stabilizer 100 to 200ppm t-butylcatechol
Formula Weight 120.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 5-Vinyl-2-norbornene, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 5-Vinyl-2-norbornene, stabilized
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.