missing translation for 'onlineSavingsMsg'
Learn More

5-Trifluoromethylphenol, TRC

Product Code. 30450124
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30450124 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450124 Supplier Toronto Research Chemicals Supplier No. TRCT791500250G
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Trifluoromethylphenol
CAS 98-17-9
Molecular Formula C7H5F3O
Quantity 250 g
Synonym 3-(Trifluoromethyl)phenol, 3-Hydroxybenzotrifluoride, NSC 9884, m-(Trifluoromethyl)phenol, m-Hydroxybenzotrifluoride, m-alpha,alpha,alpha-Trifluorocresol, alpha,alpha,alpha-Trifluoro-m-cresol
InChI Formula InChI=1S/C7H5F3O/c8-7(9,10)5-2-1-3-6(11)4-5/h1-4,11H
SMILES Oc1cccc(c1)C(F)(F)F
IUPAC Name 3-(trifluoromethyl)phenol
Molecular Weight (g/mol) 162.11
Formula Weight 162.0292
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.