missing translation for 'onlineSavingsMsg'
Learn More

5-(Trifluoromethyl)pyridine-2-carboxylic acid, 95%, Thermo Scientific Chemicals

Product Code. 15412369
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

5-(Trifluoromethyl)pyridine-2-carboxylic acid is an intermediate used in the synthesis of β-secretase (BACE) inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-(Trifluoromethyl)pyridine-2-carboxylic acid is an intermediate used in the synthesis of β-secretase (BACE) inhibitors.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80194-69-0
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.109
MDL Number MFCD04113632
InChI Key NJHGVAYLDHROPT-UHFFFAOYSA-N
Synonym 5-trifluoromethyl pyridine-2-carboxylic acid, 5-trifluoromethyl picolinic acid, 5-trifluoromethyl-2-pyridinecarboxylic acid, 5-trifluoromethyl-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 5-trifluoromethyl, 5-trifluoromethyl pyridine-2-carboxylicacid, pubchem15164, acmc-209pj2
PubChem CID 10910349
IUPAC Name 5-(trifluoromethyl)pyridine-2-carboxylic acid
SMILES C1=CC(=NC=C1C(F)(F)F)C(=O)O

Specifications

Melting Point 133°C to 137°C
Color Brown
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 191.11
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 5-(Trifluoromethyl)pyridine-2-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'