missing translation for 'onlineSavingsMsg'
Learn More

5-(Trifluoromethyl)picolinic Acid, TRC

Product Code. 30450728
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450728 50 g 50g
30449606 10 g -
30455155 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30450728 Supplier Toronto Research Chemicals Supplier No. TRCT79143050G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80194-69-0
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.11
Synonym 5-Trifluoromethylpicolinic Acid, 5-(Trifluoromethyl)-2-pyridinecarboxylic Acid
IUPAC Name 5-(trifluoromethyl)pyridine-2-carboxylic acid
SMILES OC(=O)c1ccc(cn1)C(F)(F)F

Specifications

Quantity 50 g
InChI Formula InChI=1S/C7H4F3NO2/c8-7(9,10)4-1-2-5(6(12)13)11-3-4/h1-3H,(H,12,13)
Formula Weight 191.0194
Chemical Name or Material 5-(Trifluoromethyl)picolinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.