missing translation for 'onlineSavingsMsg'
Learn More

5-(Trifluoromethyl)nicotinic acid, 97%, Thermo Scientific Chemicals

Product Code. 15440869
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15440869 1 g 1g
15450869 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15440869 Supplier Thermo Scientific Alfa Aesar Supplier No. H63906.03
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 131747-40-5 | Molecular Formula: C7H4F3NO2 | Molecular Weight: 191.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131747-40-5
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.11
MDL Number MFCD07774128
InChI Key RKFDCELCLIZRRH-UHFFFAOYSA-N
Synonym 5-trifluoromethyl nicotinic acid, 5-trifluoromethyl pyridine-3-carboxylic acid, 5-trifluoromethylinicotinic acid, 5-trifluoromethyl-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 5-trifluoromethyl, 5-trifluoromethyl-nicotinic acid, 3-pyridinecarboxylicacid, 5-trifluoromethyl, 5-trifluoromethy pyridine-3-carboxylic acid
PubChem CID 10976305
IUPAC Name 5-(trifluoromethyl)pyridine-3-carboxylic acid
SMILES OC(=O)C1=CC(=CN=C1)C(F)(F)F

Specifications

Melting Point 184°C to 189°C
Color White
Quantity 1 g
UN Number UN2811
Formula Weight 191.11
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-(Trifluoromethyl)nicotinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.