missing translation for 'onlineSavingsMsg'
Learn More

5-(Trifluoromethyl)nicotinic acid, 97%, Thermo Scientific Chemicals

Product Code. 15450869
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131747-40-5
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.11
MDL Number MFCD07774128
InChI Key RKFDCELCLIZRRH-UHFFFAOYSA-N
Synonym 5-trifluoromethyl nicotinic acid, 5-trifluoromethyl pyridine-3-carboxylic acid, 5-trifluoromethylinicotinic acid, 5-trifluoromethyl-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 5-trifluoromethyl, 5-trifluoromethyl-nicotinic acid, 3-pyridinecarboxylicacid, 5-trifluoromethyl, 5-trifluoromethy pyridine-3-carboxylic acid
PubChem CID 10976305
IUPAC Name 5-(trifluoromethyl)pyridine-3-carboxylic acid
SMILES OC(=O)C1=CC(=CN=C1)C(F)(F)F

Specifications

Melting Point 184°C to 189°C
Color White
Quantity 250 mg
UN Number UN2811
Formula Weight 191.11
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-(Trifluoromethyl)nicotinic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'