missing translation for 'onlineSavingsMsg'
Learn More

5-(Trifluoromethyl)dibenzothiophenium 1,1,1-Trifluoromethanesulfonate, TRC

Product Code. 30449051
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30449051 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449051 Supplier Toronto Research Chemicals Supplier No. TRCT791270500MG
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-(Trifluoromethyl)dibenzothiophenium 1,1,1-Trifluoromethanesulfonate
CAS 129946-88-9
Molecular Formula C14H8F6O3S2
Quantity 500 mg
Synonym Trifluoromethanesulfonic Acid Ion(1-) 5-(trifluoromethyl)dibenzothiophenium, S-(Trifluoromethyl)dibenzothiophenium Triflate, S-(Trifluoromethyl)dibenzothiophenium Trifluoromethanesulfonate
InChI Formula InChI=1S/C13H8F3S.CHF3O3S/c14-13(15,16)17-11-7-3-1-5-9(11)10-6-2-4-8-12(10)17;2-1(3,4)8(5,6)7/h1-8H;(H,5,6,7)/q+1;/p-1
SMILES [O-]S(=O)(=O)C(F)(F)F.FC(F)(F)[s+]1c2ccccc2c3ccccc13
IUPAC Name trifluoromethanesulfonate;5-(trifluoromethyl)dibenzothiophen-5-ium
Molecular Weight (g/mol) 402.33
Formula Weight 401.9819
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.