missing translation for 'onlineSavingsMsg'
Learn More

5-(Trifluoromethyl)-1H-indazole, 97%, Thermo Scientific™

Product Code. 10321420
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10321420 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10321420 Supplier Thermo Scientific Maybridge Supplier No. MAY00296DA
 Restricted Item
Estimated Shipment: 04-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 885271-64-7
Assay Percent Range 97%
Molecular Formula C8H5F3N2
Quantity 1 g
Synonym 5-trifluoromethyl-1h-indazole, 1h-indazole,5-trifluoromethyl
InChI Key WBHMPANPIGWXQV-UHFFFAOYSA-N
SMILES C1=CC2=C(C=C1C(F)(F)F)C=NN2
IUPAC Name 5-(trifluoromethyl)-1H-indazole
Molecular Weight (g/mol) 186.137
PubChem CID 21409222
Formula Weight 186.14
Chemical Name or Material 5-(Trifluoromethyl)-1H-indazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.