missing translation for 'onlineSavingsMsg'
Learn More

5-(Trifluoromethoxy)isatin, 98%

Product Code. 11467176
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11467176 5 g 5g
11457176 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11467176 Supplier Thermo Scientific Alfa Aesar Supplier No. B25494.06
  • ONLINE EXCLUSIVE PRICE
    £81.55
    £70.65 / 5g
    Save £10.90
    13% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 169037-23-4 | C9H4F3NO3 | 231.13 g/mol

5-(Trifluoromethoxy)isatin is used as a reactant for preparation of oxindole derivatives as TAk1 kinase inhibitors and of isatin thiosemicarbazones via condensation with thiosemicarbazones as herpes simplex virus (HSV) inhibitors. It is also used as a reactant for synthesis of 3-substituted 2-indolinone RET kinase inhibitors and for preparation of indolone acetamides as antiseizure agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-(Trifluoromethoxy)isatin is used as a reactant for preparation of oxindole derivatives as TAk1 kinase inhibitors and of isatin thiosemicarbazones via condensation with thiosemicarbazones as herpes simplex virus (HSV) inhibitors. It is also used as a reactant for synthesis of 3-substituted 2-indolinone RET kinase inhibitors and for preparation of indolone acetamides as antiseizure agents.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 169037-23-4
Molecular Formula C9H4F3NO3
Molecular Weight (g/mol) 231.13
MDL Number MFCD00192524
InChI Key XHAJMVPMNOBILF-UHFFFAOYSA-N
Synonym 5-trifluoromethoxy isatin, 5-trifluoromethoxy indoline-2,3-dione, 5-trifluoromethoxy-1h-indole-2,3-dione, 5-trifluoromethoxyisatin, 1h-indole-2,3-dione, 5-trifluoromethoxy, 5-trifluoromethoxy-2,3-indolinedione, buttpark 34\07-90, acmc-209dz9, d0zr2t
PubChem CID 2732752
IUPAC Name 5-(trifluoromethoxy)-1H-indole-2,3-dione
SMILES C1=CC2=C(C=C1OC(F)(F)F)C(=O)C(=O)N2

Specifications

Melting Point 170°C to 172°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 231.13
Percent Purity 98%
Chemical Name or Material 5-(Trifluoromethoxy)isatin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.