missing translation for 'onlineSavingsMsg'
Learn More

5-(Trifluoromethoxy)-1H-indole-2-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10292592
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10292592 1 g 1g
10486082 250 mg 250mg
10112934 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10292592 Supplier Thermo Scientific Maybridge Supplier No. KM09272DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175203-84-6
Molecular Formula C10H6F3NO3
Molecular Weight (g/mol) 245.16
MDL Number MFCD00276997
InChI Key DJJJSHFPEISHFN-UHFFFAOYSA-N
Synonym 5-trifluoromethoxy-1h-indole-2-carboxylic acid, 5-trifluoromethoxy indole-2-carboxylic acid, maybridge1_004876, 1h-indole-2-carboxylic acid, 5-trifluoromethoxy, 5-trifluoromethyloxy-1h-indole-2-carboxylic acid
PubChem CID 2777290
IUPAC Name 5-(trifluoromethoxy)-1H-indole-2-carboxylic acid
SMILES OC(=O)C1=CC2=CC(OC(F)(F)F)=CC=C2N1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 245.16
Percent Purity 97%
Chemical Name or Material 5-(Trifluoromethoxy)-1H-indole-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.