missing translation for 'onlineSavingsMsg'
Learn More

5-Thio-D-glucose, 97+%, Thermo Scientific Chemicals

Product Code. 15405819
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15405819 25 mg 25mg
15485809 5 mg 5mg
15495809 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15405819 Supplier Thermo Scientific Alfa Aesar Supplier No. J63621.MB
  • ONLINE EXCLUSIVE PRICE
    £133.62
    £108.65 / 25mg
    Save £24.97
    18% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 20408-97-3 | Molecular Formula: C6H12O5S | Molecular Weight: 196.217 g/mol

5-Thio-D-glucose is used as an inhibitor of the cellular transport of D-glucose and D-glucose mediated insulin release, it is also an Inhibitor of spermatogenesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Thio-D-glucose is used as an inhibitor of the cellular transport of D-glucose and D-glucose mediated insulin release, it is also an Inhibitor of spermatogenesis.

Solubility
Miscible with water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20408-97-3
Molecular Formula C6H12O5S
Molecular Weight (g/mol) 196.217
MDL Number MFCD00006663
InChI Key IJJLRUSZMLMXCN-SLPGGIOYSA-N
Synonym 5-thio-d-glucose, d-glucose, 5-thio, alpha-d-glucothiopyranose, 2r,3r,4s,5r-2,3,4,6-tetrahydroxy-5-mercaptohexanal, unii-3prv1384uo, 5-thio-alpha-d-glucopyranose, glucose, 5-thio-, d, 2r,3r,4s,5r-2,3,4,6-tetrahydroxy-5-sulfanylhexanal
PubChem CID 88527
IUPAC Name (2R,3R,4S,5R)-2,3,4,6-tetrahydroxy-5-sulfanylhexanal
SMILES C(C(C(C(C(C=O)O)O)O)S)O

Specifications

Melting Point 135°C to 138°C
Color White
Quantity 25 mg
Beilstein 1865193
Merck Index 14,9334
Solubility Information Miscible with water.
Formula Weight 196.22
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material 5-Thio-D-glucose

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.