missing translation for 'onlineSavingsMsg'
Learn More

5-(thien-2-yl)thiophene-2-carbonitrile, Thermo Scientific™

Product Code. 10203472
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10203472

Brand: Thermo Scientific Maybridge SEW02204DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 16278-99-2
Molecular Formula C9H5NS2
Molecular Weight (g/mol) 191.266
InChI Key ANARYBGZNUMARH-UHFFFAOYSA-N
Synonym 5-thien-2-yl thiophene-2-carbonitrile, 2,2'-bithiophene-5-carbonitrile, 5-thiophen-2-yl thiophene-2-carbonitrile, maybridge3_006613, 5-cyano-2,2'-bithiophene, 5-2-thienyl thiophene-2-carbonitrile
PubChem CID 2739839
IUPAC Name 5-thiophen-2-ylthiophene-2-carbonitrile
SMILES C1=CSC(=C1)C2=CC=C(S2)C#N

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 191.28
Percent Purity 97%
Chemical Name or Material 5-(Thien-2-yl)thiophene-2-carbonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.