missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(thien-2-yl)thiophene-2-carbonitrile, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Assay Percent Range | 97% |
Quantity | 250 mg |
Formula Weight | 191.28 |
Percent Purity | 97% |
Chemical Name or Material | 5-(Thien-2-yl)thiophene-2-carbonitrile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'