missing translation for 'onlineSavingsMsg'
Learn More

5-Thien-2-yl-1h-pyrazole-3-carboxylic acid, Thermo Scientific™

Product Code. 10127622
Change view
Click to view available options
Quantity:
1 g
5 g
250 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10127622 250 g 250mg
10472653 1 g 1g
10002074 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10127622 Supplier Thermo Scientific Maybridge Supplier No. CC43501CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 182415-24-3
Molecular Formula C8H6N2O2S
Molecular Weight (g/mol) 194.208
MDL Number MFCD05170066
InChI Key DUDBNCNNJMOLCC-UHFFFAOYSA-N
Synonym 5-thiophen-2-yl-1h-pyrazole-3-carboxylic acid, 5-thiophen-2-yl-2h-pyrazole-3-carboxylic acid, 3-2-thienyl-1h-pyrazole-5-carboxylic acid, 5-thien-2-yl-1h-pyrazole-3-carboxylic acid, 5-2-thienyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 5-2-thienyl, 5-2-thienyl-3-pyrazolecarboxylic acid, 3-thiophen-2-yl-1h-pyrazole-5-carboxylic acid, 5-2-thienyl pyrazole-3-carboxylic acid
PubChem CID 818268
IUPAC Name 5-thiophen-2-yl-1H-pyrazole-3-carboxylic acid
SMILES C1=CSC(=C1)C2=CC(=NN2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 g
Formula Weight 194.21
Percent Purity 97%
Chemical Name or Material 5-Thien-2-yl-1H-pyrazole-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.