missing translation for 'onlineSavingsMsg'
Learn More

5-Thiazolemethanol, TRC

Product Code. 30453141
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453141 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30453141 Supplier Toronto Research Chemicals Supplier No. TRCT34420050MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Thiazolemethanol
CAS 38585-74-9
Molecular Formula C4 H5 N O S
Quantity 50 mg
Synonym 5-Thiazolemethanol, (1,3-Thiazol-5-yl)methanol, 5-(Hydroxymethyl)-1,3-thiazole, 5-(Hydroxymethyl)thiazole, Thiazol-5-ylmethanol
InChI Formula InChI=1S/C4H5NOS/c6-2-4-1-5-3-7-4/h1,3,6H,2H2
SMILES OCc1cncs1
IUPAC Name 1,3-thiazol-5-ylmethanol
Molecular Weight (g/mol) 115.15
Formula Weight 115.0092
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.