missing translation for 'onlineSavingsMsg'
Learn More

5-Thiazolecarboxylic acid, 98%

Product Code. 15224368 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15224368 5 g 5g
15214368 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15224368 Supplier Thermo Scientific Acros Supplier No. 454540050
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14527-41-4 | C4H3NO2S | 129.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14527-41-4
Molecular Formula C4H3NO2S
Molecular Weight (g/mol) 129.13
MDL Number MFCD03428539
InChI Key YZVFSQQHQPPKNX-UHFFFAOYSA-N
Synonym thiazole-5-carboxylic acid, 5-thiazolecarboxylic acid, thiazole-5-carboxylicacid, pubchem14533, acmc-209cuh, 5-thiazole carboxylic acid, thiazole 5-carboxylic acid, ksc177g9t
PubChem CID 84494
IUPAC Name 1,3-thiazole-5-carboxylic acid
SMILES OC(=O)C1=CN=CS1

Specifications

Melting Point 208.0°C to 210.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 129.14
Percent Purity 98%
Chemical Name or Material 5-Thiazolecarboxylic acid
compliance-icons
Product Identifier
  • 5-Thiazolecarboxylic acid, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.