missing translation for 'onlineSavingsMsg'
Learn More

5-(tert-Butylsulfonyl)-2-(2-pyridyl)pyrimidin-4-amine, 97%, Thermo Scientific™

Product Code. 10582652
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10582652 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10582652 Supplier Thermo Scientific Maybridge Supplier No. KM02695EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 175202-19-4
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C13H16N4O2S
MDL Number MFCD00084869
Quantity 10 g
Synonym 5-tert-butylsulfonyl-2-2-pyridyl pyrimidin-4-amine, 5-2-methylpropane-2-sulfonyl-2-pyridin-2-yl pyrimidin-4-amine, 4-pyrimidinamine,5-1,1-dimethylethyl sulfonyl-2-2-pyridinyl, maybridge1_004567, 5-tert-butylsulfonyl-2-pyridin-2-yl pyrimidin-4-amine
InChI Key AFJQTXMEFXBCIN-UHFFFAOYSA-N
SMILES CC(C)(C)S(=O)(=O)C1=CN=C(N=C1N)C2=CC=CC=N2
IUPAC Name 5-tert-butylsulfonyl-2-pyridin-2-ylpyrimidin-4-amine
Molecular Weight (g/mol) 292.357
PubChem CID 2778673
Formula Weight 292.36
Percent Purity 97%
Chemical Name or Material 5-(tert-Butylsulfonyl)-2-(2-pyridyl)pyrimidin-4-amine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.