missing translation for 'onlineSavingsMsg'
Learn More

5-tert-Butyl-m-xylene, 98%

Product Code. 11459496
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11459496

Brand: Thermo Scientific Alfa Aesar A19220.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98-19-1 | C12H18 | 162.276 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-19-1
Molecular Formula C12H18
Molecular Weight (g/mol) 162.276
MDL Number MFCD00008832
InChI Key FZSPYHREEHYLCB-UHFFFAOYSA-N
Synonym 5-tert-butyl-m-xylene, 1-tert-butyl-3,5-dimethylbenzene, m-xylene, 5-tert-butyl, 1,3-dimethyl-5-tert-butylbenzene, benzene, 1-1,1-dimethylethyl-3,5-dimethyl, 5-tert-butyl-1,3-dimethylbenzene, benzene, 5-tert-butyl-1,3-dimethyl, 1,3-dimethyl-5-t-butylbenzene, 5-t-butyl-1,3-dimethylbenzene, benzene, 1,3-dimethyl-5-1,1-dimethylethyl
PubChem CID 7378
IUPAC Name 1-tert-butyl-3,5-dimethylbenzene
SMILES CC1=CC(=CC(=C1)C(C)(C)C)C

Specifications

Melting Point -22°C
Density 0.86
Boiling Point 207°C to 209°C
Flash Point 84°C (183°F)
Refractive Index 1.496
Quantity 500 g
Beilstein 1853314
Formula Weight 162.27
Percent Purity 98%
Chemical Name or Material 5-tert-Butyl-m-xylene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.