missing translation for 'onlineSavingsMsg'
Learn More

5-Sulphosalicylic Acid Dihydrate, Mikromol™

Product Code. 17933996
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17933996 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17933996 Supplier Mikromol Supplier No. MM0051.090025
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

5-Sulphosalicylic Acid Dihydrate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Sulphosalicylic Acid Dihydrate
CAS 5965-83-3
Analyte or Component Names 5-Sulphosalicylic Acid Dihydrate
Shipping Condition Room Temperature
Molecular Formula C7 H6 O6 S . 2 H2 O
Certifications/Compliance ISO 9001
Synonym 5-Sulphosalicylic Acid Dihydrate
InChI Formula InChI=1S/C7H6O6S.2H2O/c8-6-2-1-4(14(11,12)13)3-5(6)7(9)10;;/h1-3,8H,(H,9,10)(H,11,12,13);2*1H2
SMILES O.O.OC(=O)c1cc(ccc1O)S(=O)(=O)O
IUPAC Name 2-hydroxy-5-sulfobenzoic acid;dihydrate
Molecular Weight (g/mol) 254.21
Quantity 25 mg
Formula Weight 254.0096 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.