missing translation for 'onlineSavingsMsg'
Learn More

5-Sulfosalicylic Acid Dihydrate, TRC

Product Code. 30134095
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134095 50 g 50g
30133493 100 g 100g
17954015 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134095 Supplier Toronto Research Chemicals Supplier No. TRCS69944850G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5965-83-3
Molecular Formula C7 H6 O6 S . 2 H2 O
Molecular Weight (g/mol) 254.21
Synonym 5-Sulphosalicylic Acid Dihydrate, Benzoic acid, 2-hydroxy-5-sulfo-, hydrate (1:2), Benzoic acid, 2-hydroxy-5-sulfo-, dihydrate (9CI), Salicylic acid, 5-sulfo-, dihydrate (8CI), 5-Sulfosalicylic acid dihydrate, 2-Hydroxy-5-sulfobenzoic acid dihydrate, 3-Carboxy-4-hydroxybenzenesulfonic acid dihydrate, 5-(Hydroxysulfonyl)salicylic acid dihydrate, Salicylsulfonic acid dihydrate, Sulfosalicylic acid dihydrate, Sulphosalicylic acid dihydrate
IUPAC Name 2-hydroxy-5-sulfobenzoic acid;dihydrate
SMILES O.O.OC(=O)c1cc(ccc1O)S(=O)(=O)O

Specifications

Quantity 50 g
InChI Formula InChI=1S/C7H6O6S.2H2O/c8-6-2-1-4(14(11,12)13)3-5(6)7(9)10;;/h1-3,8H,(H,9,10)(H,11,12,13);2*1H2
Formula Weight 254.0096
Chemical Name or Material 5-Sulphosalicylic Acid Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.