missing translation for 'onlineSavingsMsg'
Learn More

5-(S-Methyl) Albendazole, TRC

Product Code. 18083740
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18083740 25 mg 25mg
18082330 10 mg 10mg
18023340 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18083740 Supplier Toronto Research Chemicals Supplier No. TRCM27195025MG
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80983-45-5
Molecular Formula C10 H11 N3 O2 S
Molecular Weight (g/mol) 237.28
Synonym Carbamic acid, [5-(methylthio)-1H-benzimidazol-2-yl]-, methyl ester (9CI), Methyl [5-(methylthio)-1H-benzimidazol-2-yl]carbamate, Methyl [5-Methylsulfanyl)-1H-benzimidazol-2-yl]carbamate, Ph Eur Albendazole Impurity F, SKF-73513
IUPAC Name methyl N-(5-methylsulfanyl-1H-benzimidazol-2-yl)carbamate
SMILES COC(=O)Nc1nc2cc(SC)ccc2[nH]1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H11N3O2S/c1-15-10(14)13-9-11-7-4-3-6(16-2)5-8(7)12-9/h3-5H,1-2H3,(H2,11,12,13,14)
Formula Weight 237.0572
Chemical Name or Material 5-(S-Methyl) Albendazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.