missing translation for 'onlineSavingsMsg'
Learn More

5-Pyrrolidin-1-ylpyridine-2-carbonitrile, 97%, Thermo Scientific™

Product Code. 10336424
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10336424 1 g Amber Glass Bottle 1g
10133174 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10336424 Supplier Thermo Scientific Maybridge Supplier No. CC52916DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 160017-09-4
Molecular Formula C10H11N3
Molecular Weight (g/mol) 173.22
MDL Number MFCD09702372
InChI Key WBEGYGZFOINKIL-UHFFFAOYSA-N
Synonym 5-pyrrolidin-1-yl pyridine-2-carbonitrile, 5-pyrrolidinylpyridine-2-carbonitrile, 2-pyridinecarbonitrile,5-1-pyrrolidinyl
PubChem CID 10261572
SMILES N#CC1=NC=C(C=C1)N1CCCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 5-(pyrrolidin-1-yl)pyridine-2-carbonitrile
Formula Weight 173.22
Percent Purity 97%
Chemical Name or Material 5-Pyrrolidin-1-ylpyridine-2-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.