missing translation for 'onlineSavingsMsg'
Learn More

(5-Pyrrolidin-1-ylpyrid-2-yl)methanol, 97%, Thermo Scientific™

Product Code. 10766905
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10766905 1 g Amber Glass Bottle 1g
10295594 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10766905 Supplier Thermo Scientific Maybridge Supplier No. CC52909DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 930110-98-8
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.24
MDL Number MFCD09702371
InChI Key GLWULQJNDPJFDR-UHFFFAOYSA-N
Synonym 5-pyrrolidin-1-ylpyrid-2-yl methanol, 5-pyrrolidin-1-ylpyridin-2-yl methanol, 5-pyrrolidin-1-yl pyridin-2-yl methanol, 5-pyrrolidinyl-2-pyridyl methan-1-ol, 5-pyrrolidine-1yl-pyrid-2-yl methanol, 2-pyridinemethanol,5-1-pyrrolidinyl, 1-6-hydroxymethyl pyridin-3-yl pyrrolidine, 2-hydroxymethyl-5-pyrrolidin-1-yl pyridine
PubChem CID 24229532
SMILES OCC1=NC=C(C=C1)N1CCCC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name [5-(pyrrolidin-1-yl)pyridin-2-yl]methanol
Formula Weight 178.23
Percent Purity 95%
Chemical Name or Material (5-Pyrrolidin-1-ylpyrid-2-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.