missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Pyrimidinylboronic acid, 97%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
£164.00 - £470.00
Chemical Identifiers
CAS | 109299-78-7 |
---|---|
Molecular Formula | C4H5BN2O2 |
Molecular Weight (g/mol) | 123.91 |
MDL Number | MFCD03002366 |
InChI Key | HZFPPBMKGYINDF-UHFFFAOYSA-N |
Synonym | 5-pyrimidinylboronic acid, pyrimidine-5-boronic acid, 5-pyrimidineboronic acid, 5-pyrimidylboronic acid, pyrimidin-5-yl boronic acid, 5-pyrimidine boronic acid, pyrimidinyl-5-boronic acid, pyrimidine-5-ylboronic acid, pyrimidin-5-yl-5-boronic acid, pyrimidin-5-boronic acid |
PubChem CID | 2795193 |
IUPAC Name | pyrimidin-5-ylboronic acid |
SMILES | OB(O)C1=CN=CN=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15204894
|
Thermo Scientific Acros
451700250 |
25 g | Glass bottle |
£164.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15224894
|
Thermo Scientific Acros
451701000 |
100 g | Glass bottle |
£470.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
109299-78-7 | |
123.91 | |
HZFPPBMKGYINDF-UHFFFAOYSA-N | |
2795193 | |
OB(O)C1=CN=CN=C1 |
C4H5BN2O2 | |
MFCD03002366 | |
5-pyrimidinylboronic acid, pyrimidine-5-boronic acid, 5-pyrimidineboronic acid, 5-pyrimidylboronic acid, pyrimidin-5-yl boronic acid, 5-pyrimidine boronic acid, pyrimidinyl-5-boronic acid, pyrimidine-5-ylboronic acid, pyrimidin-5-yl-5-boronic acid, pyrimidin-5-boronic acid | |
pyrimidin-5-ylboronic acid |
Specifications
109299-78-7 | |
96% min. (HPLC) | |
MFCD03002366 | |
HZFPPBMKGYINDF-UHFFFAOYSA-N | |
pyrimidin-5-ylboronic acid | |
2795193 | |
97% |
>300.0°C | |
C4H5BN2O2 | |
5-pyrimidinylboronic acid, pyrimidine-5-boronic acid, 5-pyrimidineboronic acid, 5-pyrimidylboronic acid, pyrimidin-5-yl boronic acid, 5-pyrimidine boronic acid, pyrimidinyl-5-boronic acid, pyrimidine-5-ylboronic acid, pyrimidin-5-yl-5-boronic acid, pyrimidin-5-boronic acid | |
OB(O)C1=CN=CN=C1 | |
123.91 | |
123.91 | |
5-Pyrimidinylboronic acid |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 5-Pyrimidinylboronic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only