missing translation for 'onlineSavingsMsg'
Learn More

5-Pyrimidinecarboxylic acid, 97%, Thermo Scientific™

Product Code. 10759195
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 4595-61-3
Molecular Formula C5H4N2O2
Molecular Weight (g/mol) 124.10
MDL Number MFCD00856162
InChI Key IIVUJUOJERNGQX-UHFFFAOYSA-N
Synonym 5-pyrimidinecarboxylic acid, 5-carboxypyrimidine, pyrimidine-5-carboxilic acid, 5-pyrimidine carboxylic acid, pyrimidine-5-carboxylate acid, pyrimidine-5-carboxylicacid, pubchem2056, acmc-209k5a, 5-pyrimidyl carboxylic acid, pyrimidin-5-carboxylic acid
PubChem CID 78346
IUPAC Name pyrimidine-5-carboxylic acid
SMILES OC(=O)C1=CN=CN=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 124.1
Percent Purity 97%
Chemical Name or Material 5-Pyrimidinecarboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'