missing translation for 'onlineSavingsMsg'
Learn More

5-Pyrid-3-ylthiophene-2-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10215474
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10215474 1 g Amber Glass Bottle 1g
10728455 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10215474 Supplier Thermo Scientific Maybridge Supplier No. CC66801DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 278803-20-6
Molecular Formula C10H7NO2S
Molecular Weight (g/mol) 205.231
MDL Number MFCD06496471
InChI Key HENDYXUZFUWIMG-UHFFFAOYSA-N
Synonym 5-pyrid-3-ylthiophene-2-carboxylic acid, 5-pyridin-3-yl thiophene-2-carboxylic acid, 2-thiophenecarboxylicacid, 5-3-pyridinyl, 5-3-pyridyl thiophene-2-carboxylic acid, 5-pyridin-3-yl-thiophene-2-carboxylic acid, 5-3-pyridinyl-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 5-3-pyridinyl
PubChem CID 4739100
IUPAC Name 5-pyridin-3-ylthiophene-2-carboxylic acid
SMILES C1=CC(=CN=C1)C2=CC=C(S2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 205.24
Percent Purity 97%
Chemical Name or Material 5-Pyrid-3-ylthiophene-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.