missing translation for 'onlineSavingsMsg'
Learn More

5-Propyl-1H-pyrazol-3-ylamine, Maybridge

Product Code. 10892801
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10892801 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10892801 Supplier Thermo Scientific Maybridge Supplier No. MAY00182DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 126748-58-1
Packaging Amber Glass Bottle
Molecular Formula C6H11N3
MDL Number MFCD08061095
Quantity 1 g
Synonym 3-amino-5-propylpyrazole, 5-propyl-1h-pyrazol-3-ylamine, 1h-pyrazol-3-amine, 5-propyl, 5-propyl-3-amino-1h-pyrazole, 3-propyl-1h-pyrazol-5-amine, 1h-pyrazol-3-amine,5-propyl-9ci, 5-propylpyrazole-3-ylamine, 3-amino-5-propyl-1h-pyrazole, 3-amino-5-n-propyl-1h-pyrazole, 5-propyl-2h-pyrazol-3-amine
InChI Key SNRYBQUHJRIMJO-UHFFFAOYSA-N
SMILES CCCC1=CC(N)=NN1
IUPAC Name 5-propyl-1H-pyrazol-3-amine
Molecular Weight (g/mol) 125.18
PubChem CID 22265684
Formula Weight 125.17
Percent Purity 97%
Chemical Name or Material 5-Propyl-1H-pyrazol-3-ylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.