missing translation for 'onlineSavingsMsg'
Learn More

5β-Pregnan-21-ol-3,11,20-trione, TRC

Product Code. 30418697
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30418697 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30418697 Supplier Toronto Research Chemicals Supplier No. TRCP71129510MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (5Beta)-21-Hydroxypregnane-3,11,20-trione
CAS 10417-86-4
Molecular Formula C21 H30 O4
Quantity 10 mg
Synonym 5Beta-Pregnan-21-ol-3,11,20-trione, Pregnane-3,11,20-trione','21-hydroxy-','(5β)-, 5β-Pregnane-3,11,20-trione','21-hydroxy- (6CI,8CI), (5β)-21-Hydroxypregnane-3,11,20-trione, 5β-Pregnan-21-ol-3,11,20-trione, DHA, NSC 79098, 5β-Dihydro-11-dehydrocorticosterone
InChI Formula InChI=1S/C21H30O4/c1-20-8-7-13(23)9-12(20)3-4-14-15-5-6-16(18(25)11-22)21(15,2)10-17(24)19(14)20/h12,14-16,19,22H,3-11H2,1-2H3/t12-,14+,15+,16-,19-,20+,21+/m1/s1
SMILES C[C@]12CCC(=O)C[C@H]1CC[C@H]3[C@@H]4CC[C@H](C(=O)CO)[C@@]4(C)CC(=O)[C@H]23
IUPAC Name (5R,8S,9S,10S,13S,14S,17S)-17-(2-hydroxyacetyl)-10,13-dimethyl-2,4,5,6,7,8,9,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthrene-3,11-dione
Molecular Weight (g/mol) 346.461
Formula Weight 346.214
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.