missing translation for 'onlineSavingsMsg'
Learn More

5β-Pregnan-21-ol-3,11,20-trione, TRC

Product Code. 30420498
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30420498 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30420498 Supplier Toronto Research Chemicals Supplier No. TRCP7112955MG
 Restricted Item
Estimated Shipment: 13-08-2026
Lägg i varukorgen
Lägg i varukorgen
Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material (5Beta)-21-Hydroxypregnane-3,11,20-trione
CAS 10417-86-4
Molecular Formula C21 H30 O4
Quantity 5 mg
Synonym 5Beta-Pregnan-21-ol-3,11,20-trione, Pregnane-3,11,20-trione','21-hydroxy-','(5β)-, 5β-Pregnane-3,11,20-trione','21-hydroxy- (6CI,8CI), (5β)-21-Hydroxypregnane-3,11,20-trione, 5β-Pregnan-21-ol-3,11,20-trione, DHA, NSC 79098, 5β-Dihydro-11-dehydrocorticosterone
InChI Formula InChI=1S/C21H30O4/c1-20-8-7-13(23)9-12(20)3-4-14-15-5-6-16(18(25)11-22)21(15,2)10-17(24)19(14)20/h12,14-16,19,22H,3-11H2,1-2H3/t12-,14+,15+,16-,19-,20+,21+/m1/s1
SMILES C[C@]12CCC(=O)C[C@H]1CC[C@H]3[C@@H]4CC[C@H](C(=O)CO)[C@@]4(C)CC(=O)[C@H]23
IUPAC Name (5R,8S,9S,10S,13S,14S,17S)-17-(2-hydroxyacetyl)-10,13-dimethyl-2,4,5,6,7,8,9,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthrene-3,11-dione
Molecular Weight (g/mol) 346.461
Formula Weight 346.214
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.