missing translation for 'onlineSavingsMsg'
Learn More

5-Phthalimidolevulinic Acid, TRC

Product Code. 30411457
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411457 2.5 g 2.5g
30412131 1 g -
30421012 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30411457 Supplier Toronto Research Chemicals Supplier No. TRCP3390152.5G
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92632-81-0
Molecular Formula C13H11NO5
Molecular Weight (g/mol) 261.23
Synonym 1,3-Dihydro-gamma,1,3-trioxo-2H-isoindole-2-pentanoic Acid, 5-(1,3-Dioxoisoindolin-2-yl)-4-oxopentanoic Acid
IUPAC Name 5-(1,3-dioxoisoindol-2-yl)-4-oxopentanoic acid
SMILES OC(=O)CCC(=O)CN1C(=O)c2ccccc2C1=O

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C13H11NO5/c15-8(5-6-11(16)17)7-14-12(18)9-3-1-2-4-10(9)13(14)19/h1-4H,5-7H2,(H,16,17)
Formula Weight 261.0637
Chemical Name or Material 5-Phthalimidolevulinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.