missing translation for 'onlineSavingsMsg'
Learn More

5-Phospho-D-ribose 1-Diphosphate Pentasodium Salt, 75%, TRC

Product Code. 30179616
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179616 5 mg 5mg
30179108 50 mg 50mg
30179289 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179616 Supplier Toronto Research Chemicals Supplier No. TRCP3537605MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108321-05-7
Molecular Formula C5H8Na5O14P3
Molecular Weight (g/mol) 499.98
Synonym 5-(Dihydrogen Phosphate) 1-(Trihydrogen Diphosphate) alpha-D-Ribofuranose Pentasodium Salt;
IUPAC Name pentasodium;[(2R,3S,4R,5R)-3,4-dihydroxy-5-[oxido(phosphonatooxy)phosphoryl]oxyoxolan-2-yl]methyl phosphate
SMILES [Na+].[Na+].[Na+].[Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](OP(=O)([O-])OP(=O)([O-])[O-])O[C@@H]1COP(=O)([O-])[O-]

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C5H13O14P3.5Na/c6-3-2(1-16-20(8,9)10)17-5(4(3)7)18-22(14,15)19-21(11,12)13;;;;;/h2-7H,1H2,(H,14,15)(H2,8,9,10)(H2,11,12,13);;;;;/q;5*+1/p-5/t2-,3-,4-,5-;;;;;/m1...../s1
Formula Weight 499.8615
Chemical Name or Material 5-Phospho-D-ribose 1-Diphosphate Pentasodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.