missing translation for 'onlineSavingsMsg'
Learn More

5-Phenyl-1H-pyrazole-4-carboxylic acid, 97%

Product Code. 15495297
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495297 250 mg 250mg
15475297 1 g 1g
15485297 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15495297 Supplier Thermo Scientific Alfa Aesar Supplier No. H32560.MD
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5504-65-4 | C10H8N2O2 | 188.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5504-65-4
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.19
MDL Number MFCD06798067,MFCD03834501
InChI Key LGTJKUYVFSBOMC-UHFFFAOYSA-N
Synonym 3-phenyl-1h-pyrazole-4-carboxylic acid, 3-phenylpyrazole-4-carboxylic acid, 3-phenyl-2h-pyrazole-4-carboxylic acid, 5-phenylpyrazole-4-carboxylic acid
PubChem CID 4138562
IUPAC Name 5-phenyl-1H-pyrazole-4-carboxylic acid
SMILES OC(=O)C1=C(NN=C1)C1=CC=CC=C1

Specifications

Melting Point 280°C to 285°C
Quantity 250 mg
Formula Weight 188.19
Percent Purity 97%
Chemical Name or Material 5-Phenyl-1H-pyrazole-4-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.