missing translation for 'onlineSavingsMsg'
Learn More

5-Phenyl-1-pentenylboronic acid pinacol ester, 96%

Product Code. 11395535
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11395535 1 g 1g
11305545 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11395535 Supplier Thermo Scientific Alfa Aesar Supplier No. L19701.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 154820-97-0 | C17H25BO2 | 272.195 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154820-97-0
Molecular Formula C17H25BO2
Molecular Weight (g/mol) 272.195
MDL Number MFCD03788752
InChI Key HLUHQMVNJDKHRO-GXDHUFHOSA-N
Synonym 5-phenyl-1-pentenylboronic acid pinacol ester, e-4,4,5,5-tetramethyl-2-5-phenylpent-1-en-1-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1e-5-phenylpent-1-en-1-yl-1,3,2-dioxaborolane
PubChem CID 10945575
IUPAC Name 4,4,5,5-tetramethyl-2-[(E)-5-phenylpent-1-enyl]-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C=CCCCC2=CC=CC=C2

Specifications

Refractive Index 1.498
Quantity 1 g
Beilstein 7295229
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 272.2
Percent Purity 96%
Chemical Name or Material 5-Phenyl-1-pentenylboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.