missing translation for 'onlineSavingsMsg'
Learn More

5-Phenyl-1-pentanol, 98%, Thermo Scientific Chemicals

Catalog Number 10180402
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
10521-91-2
C11H16O
164.25
MFCD00002979
DPZMVZIQRMVBBW-UHFFFAOYSA-N
5-phenyl-1-pentanol, benzenepentanol, 5-phenylpentanol, phenylamyl alcohol, phenylpentanol, 1-pentanol, 5-phenyl, pentanol, 5-phenyl, phenylpentanol van, unii-63ax6o7zxd, 5-phenyl-pentanol
61523
5-phenylpentan-1-ol
C1=CC=C(C=C1)CCCCCO
This item is not returnable. View return policy
10521-91-2
C11H16O
164.25
MFCD00002979
DPZMVZIQRMVBBW-UHFFFAOYSA-N
5-phenyl-1-pentanol, benzenepentanol, 5-phenylpentanol, phenylamyl alcohol, phenylpentanol, 1-pentanol, 5-phenyl, pentanol, 5-phenyl, phenylpentanol van, unii-63ax6o7zxd, 5-phenyl-pentanol
61523
5-phenylpentan-1-ol
C1=CC=C(C=C1)CCCCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 155.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5(CH2)5OH
Refractive Index 1.5162
Quantity 25 g
Beilstein 06, IV, 3373
Formula Weight 164.25
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 5-Phenyl-1-pentanol, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'