missing translation for 'onlineSavingsMsg'
Learn More

5-Phenyl-1-pentanol, 97%

Product Code. 11388595
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388595 1 g 1g
11398595 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388595 Supplier Thermo Scientific Alfa Aesar Supplier No. L12070.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 10521-91-2 | C11H16O | 164.248 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Not miscible in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10521-91-2
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.248
MDL Number MFCD00002979
InChI Key DPZMVZIQRMVBBW-UHFFFAOYSA-N
Synonym 5-phenyl-1-pentanol, benzenepentanol, 5-phenylpentanol, phenylamyl alcohol, phenylpentanol, 1-pentanol, 5-phenyl, pentanol, 5-phenyl, phenylpentanol van, unii-63ax6o7zxd, 5-phenyl-pentanol
PubChem CID 61523
IUPAC Name 5-phenylpentan-1-ol
SMILES C1=CC=C(C=C1)CCCCCO

Specifications

Density 0.975
Boiling Point 113°C to 114°C (1.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.516
Quantity 1 g
Beilstein 1935233
Solubility Information Not miscible in water.
Formula Weight 164.25
Percent Purity 97%
Chemical Name or Material 5-Phenyl-1-pentanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.