missing translation for 'onlineSavingsMsg'
Learn More

5-Phenyl-1,3-oxazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10346784
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10346784 5 g Amber Glass Bottle 5g
10561682 250 mg Amber Glass Bottle 250mg
10571682 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10346784 Supplier Thermo Scientific Maybridge Supplier No. CC10201DE
 Restricted Item
Estimated Shipment: 04-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 99924-18-2
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD00105447
InChI Key RUKDIKJSGDVSIF-UHFFFAOYSA-N
Synonym 5-phenyloxazole-4-carboxylic acid, 5-phenyl-4-oxazolecarboxylic acid, 4-oxazolecarboxylic acid, 5-phenyl, 5-phenyl-oxazole-4-carboxylic acid, buttpark 15357-35, 4-carboxy-5-phenyl-1,3-oxazole, 4-oxazolecarboxylicacid, 5-phenyl, 5-phenyl-1,3-oxazol-4-ylcarboxylic acid, saltdata: free, 2-amino-1,2,3,4-tetrahydro-5,8-dimethoxy-2-naphthalenecarboxylic acid methyl ester
PubChem CID 2776299
IUPAC Name 5-phenyl-1,3-oxazole-4-carboxylic acid
SMILES OC(=O)C1=C(OC=N1)C1=CC=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 189.17
Percent Purity 97%
Chemical Name or Material 5-Phenyl-1, 3-oxazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.