missing translation for 'onlineSavingsMsg'
Learn More

5-Phenyl-1,2,4-oxadiazol-3-amine, TRC

Product Code. 30424173
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424173 1 g 1g
30411324 100 mg -
30410747 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30424173 Supplier Toronto Research Chemicals Supplier No. TRCP4001581G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7788-14-9
Molecular Formula C8H7N3O
Molecular Weight (g/mol) 161.16
IUPAC Name 5-phenyl-1,2,4-oxadiazol-3-amine
SMILES Nc1noc(n1)c2ccccc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H7N3O/c9-8-10-7(12-11-8)6-4-2-1-3-5-6/h1-5H,(H2,9,11)
Formula Weight 161.0589
Chemical Name or Material 5-Phenyl-1,2,4-oxadiazol-3-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.