missing translation for 'onlineSavingsMsg'
Learn More

5-Oxo-1-(2-thienylmethyl)pyrrolidine-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10374382
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10374382 1 g Amber Glass Bottle 1g
10753813 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10374382 Supplier Thermo Scientific Maybridge Supplier No. DFP00358DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175136-92-2
Molecular Formula C10H11NO3S
Molecular Weight (g/mol) 225.262
MDL Number MFCD00084927
InChI Key HNEDKCTVOYJYQI-UHFFFAOYSA-N
PubChem CID 614397
IUPAC Name 5-oxo-1-(thiophen-2-ylmethyl)pyrrolidine-3-carboxylic acid
SMILES C1C(CN(C1=O)CC2=CC=CS2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 5-oxo-1-2-thienylmethyl pyrrolidine-3-carboxylic acid, 5-oxo-1-thiophen-2-ylmethyl pyrrolidine-3-carboxylic acid, 4-carboxy-1-then-2-yl pyrrolidin-2-one, 3-pyrrolidinecarboxylicacid, 5-oxo-1-2-thienylmethyl, 5-oxo-1-thien-2-ylmethyl pyrrolidine-3-carboxylic acid, maybridge1_003341, 3-pyrrolidinecarboxylicacid,5-oxo-1-2-thienylmethyl, 5-oxo-1-2-thienylmethyl-3-pyrrolidinecarboxylic acid, 5-oxo-1-2-thienylmethyl-3-pyrrolidinecarboxylic acid #, 5-oxo-1-thiophen-2-ylmethyl-pyrrolidine-3-ca rboxylic acid
Formula Weight 225.27
Percent Purity 97%
Chemical Name or Material 5-Oxo-1-(2-thienylmethyl)pyrrolidine-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.