missing translation for 'onlineSavingsMsg'
Learn More

5'-O-(tert-Butyldimethylsilyl)thymidine, 97+%, Thermo Scientific Chemicals

Product Code. 15498927
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498927 1 g 1g
15418937 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498927 Supplier Thermo Scientific Alfa Aesar Supplier No. H52301.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 40733-28-6 | Molecular Formula: C16H28N2O5Si | Molecular Weight: 356.49 g/mol

5'-O-(tert-Butyldimethylsilyl)thymidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5′-O-(tert-Butyldimethylsilyl)thymidine is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40733-28-6
Molecular Formula C16H28N2O5Si
Molecular Weight (g/mol) 356.49
MDL Number MFCD01631041
InChI Key IJWIJLIIOKZJMS-YNEHKIRRSA-N
Synonym 1-2s,4r,5s-5-tert-butyldimethylsilyl oxy methyl-4-hydroxyoxolan-2-yl-5-methyl-3h-pyrimidine-2,4-dione
PubChem CID 44717046
SMILES CC1=CN([C@H]2C[C@H](O)[C@@H](CO[Si](C)(C)C(C)(C)C)O2)C(=O)NC1=O

Specifications

Quantity 1 g
IUPAC Name 1-[(2S,4R,5S)-5-[[tert-butyl(dimethyl)silyl]oxymethyl]-4-hydroxyoxolan-2-yl]-5-methylpyrimidine-2,4-dione
Formula Weight 356.49
Percent Purity ≥97%
Chemical Name or Material 5'-O-(tert-Butyldimethylsilyl)thymidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.