missing translation for 'onlineSavingsMsg'
Learn More

5'-O-Dimethoxytrityl-deoxythymidine, 98%, Thermo Scientific™

Product Code. 10308470 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass Bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10308470 250 mg Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10308470 Supplier Thermo Scientific Acros Supplier No. 243072500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 40615-39-2
CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C31H32N2O7
MDL Number MFCD00010113
Quantity 250 mg
Synonym 5'-o-4,4'-dimethoxytrityl thymidine, dmt-t, thymidine, 5'-o-bis 4-methoxyphenyl phenylmethyl, 5'-o-4,4'-dimethoxytrityl thymidine dmt-t, 1-2r,4s,5r-5-bis 4-methoxyphenyl-phenylmethoxy methyl-4-hydroxyoxolan-2-yl-5-methylpyrimidine-2,4-dione, 5'-o-dimethoxytrityl-deoxythymidine, dmtr-dthd, dimethoxytrityl-t, 1-2r,4s,5r-5-bis 4-methoxyphenyl-phenyl-methoxy methyl-4-hydroxy-tetrahydrofuran-2-yl-5-methyl-pyrimidine-2,4-dione
InChI Key UBTJZUKVKGZHAD-QZGLRKMJNA-N
SMILES COC1=CC=C(C=C1)C(OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(C)C(=O)NC1=O)(C1=CC=CC=C1)C1=CC=C(OC)C=C1
IUPAC Name 1-[(2R,4S,5R)-5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-hydroxyoxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular Weight (g/mol) 544.60
PubChem CID 162419
Formula Weight 544.61
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5′-O-Dimethoxytrityl-deoxythymidine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.