missing translation for 'onlineSavingsMsg'
Learn More

5-O-demethyl-28-hydroxy-Avermectin A1a, TRC

Product Code. 30382596
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382596 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382596 Supplier Toronto Research Chemicals Supplier No. TRCD7220005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-O-demethyl-28-hydroxy-Avermectin A1a
CAS 96722-46-2
Molecular Formula C48 H72 O15
Quantity 5 mg
Synonym Spiro[11,15-methano-2H,13H,17H-furo[4,3,2-pq][2,6]benzodioxacyclooctadecin-, 3,2'-[2H]pyran], avermectin A1a deriv., 28-Hydroxyavermectin B1a, 8alpha-hydroxy-avermectib B1a
InChI Formula InChI=1S/C48H72O15/c1-11-24(2)42-27(5)17-18-47(63-42)23-32-20-31(62-47)16-15-26(4)41(59-38-22-36(55-10)43(30(8)57-38)60-37-21-35(54-9)40(50)29(7)56-37)25(3)13-12-14-33-45(51)61-44-39(49)28(6)19-34(46(52)58-32)48(33,44)53/h12-15,17-19,24-25,27,29-32,34-45,49-51,53H,11,16,20-23H2,1-10H3/b13-12+,26-15+,33-14-/t24-,25+,27+,29+,30+,31-,32+,34+,35+,36+,37+,38+,39-,40+,41+,42-,43+,44-,45?,47-,48-/m1/s1
SMILES CC[C@@H](C)[C@H]1O[C@@]2(C[C@@H]3C[C@@H](C\C=C(/C)\[C@@H](O[C@H]4C[C@H](OC)[C@@H](O[C@H]5C[C@H](OC)[C@@H](O)[C@H](C)O5)[C@H](C)O4)[C@@H](C)\C=C\C=C/6\C(O)O[C@@H]7[C@H](O)C(=C[C@@H](C(=O)O3)[C@]67O)C)O2)C=C[C@@H]1C
Molecular Weight (g/mol) 889.08
Formula Weight 888.4871
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.