missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-O-demethyl-28-hydroxy-Avermectin A1a, TRC
Specifications
Specifications
| Chemical Name or Material | 5-O-demethyl-28-hydroxy-Avermectin A1a |
| CAS | 96722-46-2 |
| Molecular Formula | C48 H72 O15 |
| Quantity | 1 mg |
| Synonym | Spiro[11,15-methano-2H,13H,17H-furo[4,3,2-pq][2,6]benzodioxacyclooctadecin-, 3,2'-[2H]pyran], avermectin A1a deriv., 28-Hydroxyavermectin B1a, 8alpha-hydroxy-avermectib B1a |
| InChI Formula | InChI=1S/C48H72O15/c1-11-24(2)42-27(5)17-18-47(63-42)23-32-20-31(62-47)16-15-26(4)41(59-38-22-36(55-10)43(30(8)57-38)60-37-21-35(54-9)40(50)29(7)56-37)25(3)13-12-14-33-45(51)61-44-39(49)28(6)19-34(46(52)58-32)48(33,44)53/h12-15,17-19,24-25,27,29-32,34-45,49-51,53H,11,16,20-23H2,1-10H3/b13-12+,26-15+,33-14-/t24-,25+,27+,29+,30+,31-,32+,34+,35+,36+,37+,38+,39-,40+,41+,42-,43+,44-,45?,47-,48-/m1/s1 |
| SMILES | CC[C@@H](C)[C@H]1O[C@@]2(C[C@@H]3C[C@@H](C\C=C(/C)\[C@@H](O[C@H]4C[C@H](OC)[C@@H](O[C@H]5C[C@H](OC)[C@@H](O)[C@H](C)O5)[C@H](C)O4)[C@@H](C)\C=C\C=C/6\C(O)O[C@@H]7[C@H](O)C(=C[C@@H](C(=O)O3)[C@]67O)C)O2)C=C[C@@H]1C |
| Molecular Weight (g/mol) | 889.08 |
| Formula Weight | 888.4871 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?